ENA423209512 Enamine

N-cyclopentyl-4-nitrobenzene-1-sulfonamide

  • MDL number MFCD00451512
  • Cas Number : 413573-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170257

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-170257
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart