ENA423209514 Enamine

cyclopentylmethyl methanesulfonate

  • MDL number MFCD26961254
  • Cas Number : 73017-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170262

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170262
Availability details for
Quantity
  • Availability
  • Add to Cart