ENA423209519 Enamine

2-[(2-hydroxypropyl)amino]propan-1-ol

  • MDL number MFCD12801445
  • Cas Number : 115893-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170294

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-170294
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart