ENA423209521 Enamine

7,7-dimethyl-6-oxa-9-azaspiro[4.5]decane

  • MDL number MFCD21126222
  • Cas Number : 1488879-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170297
Availability details for
Quantity
  • Availability
  • Add to Cart