ENA423209523 Enamine

3,5,6-trichloropyridin-2-amine

  • MDL number MFCD11847442
  • Cas Number : 1378670-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170299
Availability details for
Quantity
  • Availability
  • Add to Cart