ENA423209529 Enamine

2-[6-chloro-4-(trifluoromethyl)pyridin-2-yl]acetic acid

  • MDL number MFCD22546213
  • Cas Number : 1393573-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170319
Availability details for
Quantity
  • Availability
  • Add to Cart