ENA423209530 Enamine

3-(trifluoromethyl)pyrazin-2-amine

  • MDL number MFCD15144417
  • Cas Number : 213019-67-1
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(trifluoromethyl)-2-pyrazinamine
Inchi Code 1S/C5H4F3N3/c6-5(7,8)3-4(9)11-2-1-10-3/h1-2H,(H2,9,11)
Inchi Key GDOQHWXDKQJKLT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170321

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-170321
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart