ENA423209532 Enamine

{4-[2-(2-methoxyethoxy)ethoxy]phenyl}methanol

  • MDL number MFCD19672593
  • Cas Number : 392711-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170327
Availability details for
Quantity
  • Availability
  • Add to Cart