ENA423209533 Enamine

4-[2-(2-methoxyethoxy)ethoxy]benzaldehyde

  • MDL number MFCD19672512
  • Cas Number : 64994-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170328

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170328
Availability details for
Quantity
  • Availability
  • Add to Cart