ENA423209537 Enamine

(4-acetyl-2-fluorophenyl)boronic acid

  • MDL number MFCD22570911
  • Cas Number : 1022154-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170360
Availability details for
Quantity
  • Availability
  • Add to Cart