ENA423209541 Enamine

3-(2-bromophenyl)-5-(pyrrolidin-2-yl)-1,2,4-oxadiazole

  • MDL number MFCD16506273
  • Cas Number : 1285394-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170364
Availability details for
Quantity
  • Availability
  • Add to Cart