ENA423209542 Enamine

1-bromo-5-(bromomethyl)naphthalene

  • MDL number MFCD16249711
  • Cas Number : 151109-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170366
Availability details for
Quantity
  • Availability
  • Add to Cart