ENA423209544 Enamine

3-(1-bromoethyl)benzonitrile

  • MDL number MFCD19625763
  • Cas Number : 1096159-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170368

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170368
Availability details for
Quantity
  • Availability
  • Add to Cart