ENA423209556 Enamine

1,2,3,4,5,6,7,8-octahydroquinoline-2,5-dione

  • MDL number MFCD01111799

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170398
Availability details for
Quantity
  • Availability
  • Add to Cart