ENA423209567 Enamine

2-amino-2-(4-methyl-1,3-thiazol-5-yl)acetic acid

  • MDL number MFCD20488188
  • Cas Number : 50382-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170413
Availability details for
Quantity
  • Availability
  • Add to Cart