ENA423209596 Enamine

methyl 3-(3,3-difluorocyclobutyl)-3-oxopropanoate

  • MDL number MFCD17015923
  • Cas Number : 1191096-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170510
Availability details for
Quantity
  • Availability
  • Add to Cart