ENA423209601 Enamine

3-bromo-5-fluoro-2-hydroxybenzoic acid

  • MDL number MFCD11203628
  • Cas Number : 4180-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170530
Availability details for
Quantity
  • Availability
  • Add to Cart