ENA423209603 Enamine

1-{[(9H-fluoren-9-yl)methoxy]carbonyl}azepane-2-carboxylic acid

  • MDL number MFCD20362135
  • Cas Number : 632350-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170533
Availability details for
Quantity
  • Availability
  • Add to Cart