ENA423209604 Enamine

1-methyl-1H-indol-7-amine

  • MDL number MFCD18377766
  • Cas Number : 102308-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170535

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-170535
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart