ENA423209608 Enamine

2-(2-ethylphenyl)acetic acid

  • MDL number MFCD09924863
  • Cas Number : 19418-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170550

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170550
Availability details for
Quantity
  • Availability
  • Add to Cart