ENA423209611 Enamine

5-(trifluoromethyl)quinoline

  • MDL number MFCD16619721
  • Cas Number : 342-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170554

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-170554
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart