ENA423209619 Enamine

2-(1,4-diazepan-1-yl)propan-1-ol

  • MDL number MFCD12781496
  • Cas Number : 1184691-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170565
Availability details for
Quantity
  • Availability
  • Add to Cart