ENA423209620 Enamine

cyano(methyl)amine

  • MDL number MFCD16767882
  • Cas Number : 4674-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170566

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-170566
pictograms GHS01,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart