ENA423209621 Enamine

5-(5-methyl-1-benzofuran-2-yl)-1,3,4-oxadiazol-2-amine

  • Cas Number : 1248746-00-0
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(5-methyl-1-benzofuran-2-yl)-1,3,4-oxadiazol-2-amine
Inchi Code 1S/C11H9N3O2/c1-6-2-3-8-7(4-6)5-9(15-8)10-13-14-11(12)16-10/h2-5H,1H3,(H2,12,14)
Inchi Key TZBHUDJZYWMQOX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170569
Availability details for
Quantity
  • Availability
  • Add to Cart