ENA423209637 Enamine

1-(5-fluoro-1-benzofuran-2-yl)ethan-1-amine

  • MDL number MFCD21234003
  • Cas Number : 159009-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170722
Availability details for
Quantity
  • Availability
  • Add to Cart