ENA423209646 Enamine

2,2-diethyl-4-methoxy-4-oxobutanoic acid

  • MDL number MFCD20643067
  • Cas Number : 90647-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170757

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170757
Availability details for
Quantity
  • Availability
  • Add to Cart