ENA423209647 Enamine

2-chloro-6-(propan-2-ylsulfanyl)benzaldehyde

  • MDL number MFCD18887924
  • Cas Number : 1341438-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170759

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170759
Availability details for
Quantity
  • Availability
  • Add to Cart