ENA423209649 Enamine

1-(bromomethyl)-3-chloro-2-fluorobenzene

  • MDL number MFCD00042503
  • Cas Number : 85070-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30...32
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170762

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-170762
pictograms GHS05,GHS07,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart