ENA423209652 Enamine

6-bromo-3,5-dichloropyridin-2-amine

  • MDL number MFCD11044254
  • Cas Number : 105751-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170766
Availability details for
Quantity
  • Availability
  • Add to Cart