ENA423209654 Enamine

{[3-bromo-2-(oxolan-2-ylmethoxy)phenyl]methyl}(methyl)amine

  • MDL number MFCD13479381
  • Cas Number : 1291714-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170768

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170768
Availability details for
Quantity
  • Availability
  • Add to Cart