ENA423209655 Enamine

4-methoxy-N-(piperidin-4-yl)pyrimidin-2-amine

  • MDL number MFCD12086997
  • Cas Number : 1156837-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170769

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170769
Availability details for
Quantity
  • Availability
  • Add to Cart