ENA423209658 Enamine

3,4,5-trichloropyridin-2-amine

  • MDL number MFCD08705655
  • Cas Number : 55933-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170774
Availability details for
Quantity
  • Availability
  • Add to Cart