ENA423209666 Enamine

N-(azetidin-3-yl)acetamide hydrochloride

  • MDL number MFCD08059477
  • Cas Number : 102065-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170805
Availability details for
Quantity
  • Availability
  • Add to Cart