ENA423209674 Enamine

6-methoxy-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD00128534
  • Cas Number : 13735-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170867
Availability details for
Quantity
  • Availability
  • Add to Cart