ENA423209675 Enamine

2-(1-methyl-1H-pyrazol-4-yl)pyridin-3-amine

  • MDL number MFCD24254905
  • Cas Number : 1554842-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170868
Availability details for
Quantity
  • Availability
  • Add to Cart