ENA423209678 Enamine

1-methyl-7-nitro-1H-indole

  • MDL number MFCD23378869
  • Cas Number : 101489-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170873
Availability details for
Quantity
  • Availability
  • Add to Cart