ENA423209680 Enamine

methyl 2,2-dimethyl-3-(propylsulfanyl)propanoate

  • MDL number MFCD23731761
  • Cas Number : 1502982-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170877
Availability details for
Quantity
  • Availability
  • Add to Cart