ENA423209687 Enamine

quinoline-4-carbonitrile

  • MDL number MFCD00972111
  • Cas Number : 2973-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170895
Availability details for
Quantity
  • Availability
  • Add to Cart