ENA423209693 Enamine

octahydrocyclopenta[b][1,4]thiazine

  • MDL number MFCD19159789
  • Cas Number : 30196-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170904
Availability details for
Quantity
  • Availability
  • Add to Cart