ENA423209699 Enamine

4-[(3-fluoro-4-nitrophenyl)methyl]morpholine

  • MDL number MFCD07368797
  • Cas Number : 552883-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171028

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171028
Availability details for
Quantity
  • Availability
  • Add to Cart