ENA423209700 Enamine

2-(cyclopentylmethyl)-5-iodo-1,3,4-thiadiazole

  • MDL number MFCD18345376
  • Cas Number : 1339636-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171030
Availability details for
Quantity
  • Availability
  • Add to Cart