ENA423209704 Enamine

2-(3,4-dichlorophenoxy)-N-methylpropanamide

  • MDL number MFCD27980891
  • Cas Number : 1623912-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171035
Availability details for
Quantity
  • Availability
  • Add to Cart