ENA423209705 Enamine

methyl 2-(3,4-dichlorophenoxy)propanoate

  • MDL number MFCD03942994
  • Cas Number : 63461-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171036

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171036
Availability details for
Quantity
  • Availability
  • Add to Cart