ENA423209708 Enamine

ethyl 2-(3,4-dichlorophenoxy)butanoate

  • MDL number MFCD20540806
  • Cas Number : 1378309-19-3
  • Molecular Weight : 277.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171040
Availability details for
Quantity
  • Availability
  • Add to Cart