ENA423209713 Enamine

3-methyl-3-[4-(trifluoromethyl)phenyl]pyrrolidine

  • MDL number MFCD16737841
  • Cas Number : 1248089-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171051
Availability details for
Quantity
  • Availability
  • Add to Cart