ENA423209714 Enamine

4-amino-2-(ethylsulfamoyl)-N,N-dimethylbenzamide

  • MDL number MFCD26141962
  • Cas Number : 1803593-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171055
Availability details for
Quantity
  • Availability
  • Add to Cart