ENA423209736 Enamine

4-(3-bromobenzoyl)oxane

  • MDL number MFCD19611767
  • Cas Number : 1339838-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171148
Availability details for
Quantity
  • Availability
  • Add to Cart