ENA423209741 Enamine

2-(trifluoromethyl)-1,3-thiazole-4-carbonyl chloride

  • MDL number MFCD11558916
  • Cas Number : 1076197-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171157
Availability details for
Quantity
  • Availability
  • Add to Cart