ENA423209754 Enamine

2-bromo-4-iodo-1-(trifluoromethyl)benzene

  • MDL number MFCD22491025
  • Cas Number : 1369927-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-171217
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart