ENA423209755 Enamine

(2-isothiocyanatopropyl)benzene

  • MDL number MFCD00126014
  • Cas Number : 16220-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171218
Availability details for
Quantity
  • Availability
  • Add to Cart